Bis(1,2,2,6,6-pentamethyl-4-piperidyl)sebacate ≥95.0%
Leverantör: TCI EUROPE
Varning
Synonyms: Bis(1,2,2,6,6-pentamethylpiperidin-4-yl) decanedioate, Decanedioic Acid Bis(1,2,2,6,6-pentamethyl-4-piperidyl) Ester, Sebacic Acid Bis(1,2,2,6,6-pentamethyl-4-piperidyl) Ester
B3924-5G B3924-25G
TCIAB3924-5GEA 535 SEK
TCIAB3924-5G TCIAB3924-25G
Bis(1,2,2,6,6-pentamethyl-4-piperidyl)sebacate ≥95.0%
Bis(1,2,2,6,6-pentamethyl-4-piperidyl)sebacate
Formel: C₃₀H₅₆N₂O₄
MW: 508,79 g/mol
Kokpunkt: 222°C
MDL Number: MFCD00134706
CAS nummer: 41556-26-7
EINECS: 255-437-1
Order Now
Specifikation testresultat
Appearance White to Almost white clear liquid
Purity (GC) min. 95.0 %