(S)-2-Amino-4-(tert-butoxy)-4-oxobutanoic acid ≥98.0%
Leverantör: TCI EUROPE
Synonyms:
4-tert-Butyl L-Aspartate, H-Asp(OtBu)-OH, L-Aspartic acid 4-tert-butyl ester, L-Aspartic acid 4-tert-Butyl ester
B3919-1G
B3919-5G
TCIAB3919-1GEA
630
SEK
TCIAB3919-1G
TCIAB3919-5G
(S)-2-Amino-4-(tert-butoxy)-4-oxobutanoic acid ≥98.0%
(S)-2-Amino-4-(tert-butoxy)-4-oxobutanoic acid
Formel:
C₈H₁₅NO₄ MW: 189,2 g/mol |
MDL Number:
MFCD00038577 CAS nummer: 3057-74-7 EINECS: 221-292-8 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Specific rotation [a]20/D | +8.0 to +10.0 deg (C=1, H2O) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...