Bromomethyl acetate ≥95.0%

Leverantör: TCI EUROPE
Varning

B3907-1G B3907-5G
TCIAB3907-1GEA 615 SEK
TCIAB3907-1G TCIAB3907-5G
Bromomethyl acetate ≥95.0%
Bromomethyl acetate
Formel: H₃CCOOCH₂Br
MW: 152,98 g/mol
Kokpunkt: 134°C
Flampunkt: 57°C
Storage Temperature: Kylskåp
MDL Number: MFCD00000170
CAS nummer: 590-97-6
EINECS: 209-696-2
UN: 3272
ADR: 3,III

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR