2-Bromo-9,9-dimethylfluorene ≥98.0%

Leverantör: TCI EUROPE
Varning

B3894-5G B3894-25G
TCIAB3894-5GEA 310 SEK
TCIAB3894-5G TCIAB3894-25G
2-Bromo-9,9-dimethylfluorene ≥98.0%
2-Bromo-9,9-dimethylfluorene
Formel: C₁₅H₁₃Br
MW: 273,17 g/mol
Kokpunkt: 190°C
MDL Number: MFCD09261272
CAS nummer: 28320-31-2

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 67.0 to 71.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR