4-Bromo-1-indanone ≥97.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 4-bromo-2,3-dihydro-1H-inden-1-one

B3801-1G B3801-5G
TCIAB3801-1GEA 375 SEK
TCIAB3801-1G TCIAB3801-5G
4-Bromo-1-indanone ≥97.0%
4-Bromo-1-indanone
Formel: C₉H₇BrO
MW: 211,06 g/mol
Kokpunkt: 125°C
MDL Number: MFCD01719772
CAS nummer: 15115-60-3

Order Now

Specifikation testresultat

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 97.0 %
Melting point 92.0 to 96.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR