2,2'-Bibenzo[d][1,3,2]dioxaborole ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: Bis(catecholato)diborane

B3757-1G B3757-5G
TCIAB3757-1GEA 510 SEK
TCIAB3757-1G TCIAB3757-5G
2,2'-Bibenzo[d][1,3,2]dioxaborole ≥98.0%
2,2'-Bibenzo[d][1,3,2]dioxaborole
Formel: C₁₂H₈B₂O₄
MW: 237,8115 g/mol
MDL Number: MFCD09842343
CAS nummer: 13826-27-2

Order Now

Specifikation testresultat

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 194.0 to 198.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR