Benzydamine hydrochloride ≥97.0% (by HPLC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
Benzydamine HCl, 1-Benzyl-3-[3-(dimethylamino)propoxy]-1H-indazole Hydrochloride, 3-((1-Benzyl-1H-indazol-3-yl)oxy)-N,N-dimethylpropan-1-amine hydrochloride
B3751-5G
B3751-25G
TCIAB3751-5GEA
495
SEK
TCIAB3751-5G
TCIAB3751-25G
Benzydamine hydrochloride ≥97.0% (by HPLC, titration analysis)
Benzydamine hydrochloride
Formel:
C₁₉H₂₃N₃O MW: 345,87 g/mol Storage Temperature: Kylskåp |
MDL Number:
MFCD00078957 CAS nummer: 132-69-4 EINECS: 205-076-0 Merck Index: 14,01122 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (Argentmetric Titration) | min. 97.0 % |
Melting point | 157.0 to 162.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...