2-Bromo-5-n-octylthiophene ≥97.0%

Leverantör: TCI EUROPE

B3738-25G B3738-5G
TCIAB3738-25GEA 6750 SEK
TCIAB3738-25G TCIAB3738-5G
2-Bromo-5-n-octylthiophene ≥97.0%
2-Bromo-5-n-octylthiophene
Formel: C₁₂H₁₉BrS
MW: 275,25 g/mol
Kokpunkt: 112°C
Smältpunkt: 5°C
Storage Temperature: Kylskåp
CAS nummer: 172514-63-5

Order Now

Specifikation testresultat

Appearance Colorless to Yellow to Orange clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR