5-Bromoresorcinol ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: 1-Bromo-3,5-dihydroxybenzene, 5-Bromobenzene-1,3-diol, 3,5-Dihydroxybromobenzene

B3731-1G B3731-5G
TCIAB3731-1GEA 790 SEK
TCIAB3731-1G TCIAB3731-5G
5-Bromoresorcinol ≥98.0% (by GC, titration analysis)
5-Bromoresorcinol
Formel: C₆H₅BrO₂
MW: 189,01 g/mol
Kokpunkt: 140°C
MDL Number: MFCD16293393
CAS nummer: 106120-04-1

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Bromination Method) min. 98.0 %
Melting point 90.0 to 96.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR