2-(tert-Butylamino)-1-(3-chlorophenyl)propan-1-one hydrochloride ≥98.0% (by HPLC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: Amfebutamone hydrochloride, Bupropion Hydrochloride, Bupropion hydrochloride, 2-(tert-Butylamino)-1-(3-chlorophenyl)-1-propanone hydrochloride

B3649-1G B3649-5G
TCIAB3649-1GEA 575 SEK
TCIAB3649-1G TCIAB3649-5G
2-(tert-Butylamino)-1-(3-chlorophenyl)propan-1-one hydrochloride ≥98.0% (by HPLC, titration analysis)
2-(tert-Butylamino)-1-(3-chlorophenyl)propan-1-one hydrochloride
Formel: C₁₃H₁₉Cl₂NO
MW: 276,20 g/mol
MDL Number: MFCD00055209
CAS nummer: 31677-93-7
EINECS: 250-759-9
Merck Index: 14,01499

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR