Buflomedil hydrochloride ≥98.0% (by HPLC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
4-(Pyrrolidin-1-yl)-1-(2,4,6-trimethoxyphenyl)butan-1-one hydrochloride, 4-(1-Pyrrolidinyl)-1-(2,4,6-trimethoxyphenyl)-1-butanone Hydrochloride, Buflomedil HCl
B3647-5G
B3647-1G
TCIAB3647-5GEA
1325
SEK
TCIAB3647-5G
TCIAB3647-1G
Buflomedil hydrochloride ≥98.0% (by HPLC, titration analysis)
Buflomedil hydrochloride
Formel:
C₁₇H₂₅NO₄ MW: 343,85 g/mol Smältpunkt: 193°C |
MDL Number:
MFCD00078965 CAS nummer: 35543-24-9 EINECS: 252-611-9 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...