N,N'-Bis-Z-1-guanylpyrazole ≥98.0% (by HPLC, titration analysis)

Leverantör: TCI
Varning

Synonyms: N,N'-Bis-Z-1-Guanylpyrazole

B3605-1G B3605-5G
TCIAB3605-1GEA 870 SEK
TCIAB3605-1G TCIAB3605-5G
N,N'-Bis-Z-1-guanylpyrazole ≥98.0% (by HPLC, titration analysis)
N,N'-Bis-Z-1-guanylpyrazole
Formel: C₂₀H₁₈N₄O₄
MW: 378.38 g/mol
MDL Number: MFCD02683516
CAS nummer: 152120-55-3

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 91.0 to 95.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR