3-Benzoylthiazolidine-2-thione ≥98.0%

Leverantör: TCI EUROPE

B3571-1G B3571-5G
TCIAB3571-1GEA 805 SEK
TCIAB3571-1G TCIAB3571-5G
3-Benzoylthiazolidine-2-thione ≥98.0%
3-Benzoylthiazolidine-2-thione
Formel: C₁₀H₉NOS₂
MW: 223,31 g/mol
CAS nummer: 70326-37-3

Order Now

Specifikation testresultat

Appearance Light yellow to Yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 108.0 to 112.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR