Vismut(III)klorid, vattenfri ≥97.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
Bismuth(III) chloride, Bismuth trichloride
B3546-250G
B3546-25G
TCIAB3546-250GEA
2450
SEK
TCIAB3546-250G
TCIAB3546-25G
Vismut(III)klorid, vattenfri ≥97.0%
Vismut(III)klorid
Formel:
BiCl₃ MW: 315,34 g/mol Kokpunkt: 447°C Smältpunkt: 230°C |
MDL Number:
MFCD00003461 CAS nummer: 7787-60-2 EINECS: 232-123-2 Merck Index: 14,01262 |
Specifikation testresultat
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (Chelometric Titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...