Bezafibrate ≥98.0% (by HPLC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: 2-(4-(2-(4-Chlorobenzamido)ethyl)phenoxy)-2-methylpropanoic acid, 2-[4-[2-(4-Chlorobenzamido)ethyl]phenoxy]isobutyric Acid

B3346-25G B3346-5G
TCIAB3346-25GEA 3325 SEK
TCIAB3346-25G TCIAB3346-5G
Bezafibrate ≥98.0% (by HPLC, titration analysis)
Bezafibrate
Formel: C₁₉H₂₀ClNO₄
MW: 361,82 g/mol
MDL Number: MFCD00078970
CAS nummer: 41859-67-0
EINECS: 255-567-9
Merck Index: 14,01195

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 182.0 to 186.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR