[(tert-Butoxycarbonyl)aminooxy]acetic acid ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: N-Boc-(carboxymethoxy)amine, Boc-Aoa-OH, (Boc-aminooxy)acetic Acid

B3338-1G B3338-5G
TCIAB3338-1GEA 615 SEK
TCIAB3338-1G TCIAB3338-5G
[(tert-Butoxycarbonyl)aminooxy]acetic acid ≥98.0%
[(tert-Butoxycarbonyl)aminooxy]acetic acid
Formel: C₇H₁₃NO₅
MW: 191,18 g/mol
MDL Number: MFCD01632027
CAS nummer: 42989-85-5

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR