9-Bromo-1-nonene ≥95.0%
Leverantör: TCI EUROPE
Varning
B3256-1G B3256-5G
TCIAB3256-1GEA 335 SEK
TCIAB3256-1G TCIAB3256-5G
9-Bromo-1-nonene ≥95.0%
9-Bromo-1-nonene
Formel: C₉H₁₇Br
MW: 205,139 g/mol
Kokpunkt: 101°C
Storage Temperature: Kylskåp
MDL Number: MFCD09037826
CAS nummer: 89359-54-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1f723990cdf5ef33d816ad150e10310372496dcd&vid=1f723990cdf5ef33d816ad150e10310372496dcd&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %