Benzyl-2,2,2-Trifluoro-N-phenylacetimidate ≥98.0%
Leverantör: TCI
Varning
Synonyms: 2,2,2-Trifluoro-N-phenylacetimidic Acid Benzyl Ester
B3234-5G B3234-1G
TCIAB3234-5GEA 6525 SEK
TCIAB3234-5G TCIAB3234-1G
Benzyl-2,2,2-Trifluoro-N-phenylacetimidate ≥98.0%
Benzyl-2,2,2-Trifluoro-N-phenylacetimidate
Formel: C₁₅H₁₂F₃NO
MW: 279.26 g/mol
Storage Temperature: Kylskåp
CAS nummer: 952057-61-3
Order Now
Specifikation testresultat
Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %