3-Bromothieno[3,2-b]thiophene ≥94.0%
Leverantör: TCI EUROPE
B3226-1G B3226-200MG
TCIAB3226-1GEA 6575 SEK
TCIAB3226-1G TCIAB3226-200MG
3-Bromothieno[3,2-b]thiophene ≥94.0%
3-Bromothieno[3,2-b]thiophene
Formel: C₆H₃BrS₂
MW: 219,11 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD10000952
CAS nummer: 25121-83-9
Order Now
Specifikation testresultat
Appearance Colorless to Yellow clear liquid
Purity (GC) min. 94.0 %