4'-Bromo-2,2':6',2''-terpyridine ≥97.0% (by HPLC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
4'-Bromo-alpha,alpha',alpha''-tripyridyl
B3219-1G
B3219-5G
TCIAB3219-1GEA
3725
SEK
TCIAB3219-1G
TCIAB3219-5G
4'-Bromo-2,2':6',2''-terpyridine ≥97.0% (by HPLC, titration analysis)
4'-Bromo-2,2':6',2''-terpyridine
Formel:
C₁₅H₁₀BrN₃ MW: 312,17 g/mol |
MDL Number:
MFCD06796965 CAS nummer: 149817-62-9 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (Nonaqueous Titration) | min. 97.0 % |
Melting point | 135.0 to 140.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...