4-(N-Butyl-N-nitrosamino)butyric acid ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: BCPN, N-Butyl-N-(3-carboxypropyl)nitrosamine

B3185-1G
TCIAB3185-1GEA 2800 SEK
TCIAB3185-1G
4-(N-Butyl-N-nitrosamino)butyric acid ≥98.0%
4-(N-Butyl-N-nitrosamino)butyric acid
Formel: C₈H₁₆N₂O₃
MW: 188,23 g/mol
Storage Temperature: Kylskåp
CAS nummer: 38252-74-3

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (GC) min. 96.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 38.0 and le 43.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR