4-(4-Bromophenyl)butyric acid ≥98.0%

Leverantör: TCI
Varning

B3131-5G B3131-1G
TCIAB3131-5GEA 1300 SEK
TCIAB3131-5G TCIAB3131-1G
4-(4-Bromophenyl)butyric acid ≥98.0%
4-(4-Bromophenyl)butyric acid
Formel: C₁₀H₁₁BrO₂
Kokpunkt: 176°C
MDL Number: MFCD00463170
CAS nummer: 35656-89-4

Order Now

Specifikation testresultat

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 69.0 to 73.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR