N-Benzylglycine hydrochloride ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: Bzl-Gly-OH·HCl, Bzl-Gly-OH.HCl

B3075-25G B3075-5G
TCIAB3075-25GEA 1425 SEK
TCIAB3075-25G TCIAB3075-5G
N-Benzylglycine hydrochloride ≥98.0%
N-Benzylglycine hydrochloride
Formel: C₉H₁₁NO₂·HCl
MW: 201,7 g/mol
Smältpunkt: 228°C
MDL Number: MFCD00156934
CAS nummer: 7689-50-1

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR