1-Benzyl-L-glutamate ≥98.0% (by HPLC, titration analysis)

Leverantör: TCI EUROPE

Synonyms: L-Glutamic Acid 1-Benzyl Ester, H-Glu-OBzl, (S)-4-Amino-5-(benzyloxy)-5-oxopentanoic acid

B2996-1G B2996-5G
TCIAB2996-1GEA 365 SEK
TCIAB2996-1G TCIAB2996-5G
1-Benzyl-L-glutamate ≥98.0% (by HPLC, titration analysis)
1-Benzyl-L-glutamate
Formel: C₁₂H₁₅NO₄
MW: 237,26 g/mol
MDL Number: MFCD00037312
CAS nummer: 13030-09-6

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D +10.0 to +12.0 deg (C=1, 1mol/L HCl)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR