1,2-Bis(phenylthio)ethylene (cis and trans mixture) cis- and trans-mixture ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
1,2-Bis(phenylmercapto)ethylene
B2940-25G
B2940-5G
TCIAB2940-25GEA
2825
SEK
TCIAB2940-25G
TCIAB2940-5G
1,2-Bis(phenylthio)ethylene (cis and trans mixture) cis- and trans-mixture ≥98.0%
1,2-Bis(phenylthio)ethylene (cis and trans mixture)
Formel:
C₁₄H₁₂S₂ MW: 244,37 g/mol Kokpunkt: 166°C Smältpunkt: 48°C Storage Temperature: Kylskåp |
CAS nummer:
23528-44-1 |
Specifikation testresultat
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...