(S)-2-Methylpropane-2-sulphinamide ≥98.0%

Leverantör: TCI EUROPE

Synonyms: (S)-tert-Butylsulphinamide, (S)-(-)-tert-Butylsulphinamide, (S)-(-)-t-Butylsulphinamide, (S)-(-)-2-Methyl-2-propanesulphinamide, (S)-(-)-tert-Butyl sulphinamide

B2908-1G B2908-5G
TCIAB2908-1GEA 285 SEK
TCIAB2908-1G TCIAB2908-5G
(S)-2-Methylpropane-2-sulphinamide ≥98.0%
(S)-2-Methylpropane-2-sulphinamide
Formel: C₄H₁₁NOS
MW: 121,20 g/mol
MDL Number: MFCD05861480
CAS nummer: 343338-28-3

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 103.0 to 108.0 deg-C
Specific rotation [a]20/D -2.0 to -6.0 deg (C=1, CHCl3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR