(R)-(+)-2-Methyl-2-propanesulphinamide ≥98.0%

Leverantör: TCI EUROPE

Synonyms: (R)-2-methylpropane-2-sulphinamide, R)-(+)-tert-Butyl sulphinamide, (R)-2-Methyl-2-propanesulphinamide, (R)-(+)-tert-Butylsulphinamide, (R)-(+)-t-Butylsulphinamide

B2907-1G B2907-5G
TCIAB2907-1GEA 335 SEK
TCIAB2907-1G TCIAB2907-5G
(R)-(+)-2-Methyl-2-propanesulphinamide ≥98.0%
(R)-(+)-2-Methyl-2-propanesulphinamide
Formel: C₄H₁₁NOS
MW: 121,20 g/mol
MDL Number: MFCD05861479
CAS nummer: 196929-78-9

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 104.0 to 108.0 deg-C
Specific rotation [a]20/D +2.0 to +6.0 deg (C=1, CHCl3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR