2-(2-Bromophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97.0%

Leverantör: TCI EUROPE

Synonyms: 2-Bromophenylboronic Acid Pinacol Ester, 1-Bromo-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene

B2828-1G B2828-5G
TCIAB2828-1GEA 1950 SEK
TCIAB2828-1G TCIAB2828-5G
2-(2-Bromophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97.0%
2-(2-Bromophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formel: C₁₂H₁₆BBrO₂
MW: 282,97 g/mol
MDL Number: MFCD08276318
CAS nummer: 269410-06-2

Order Now

Specifikation testresultat

Appearance Colorless to Light orange to Yellow clear liquid
Purity (Neutralization titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR