((3-Bromopropoxy)methyl)benzene ≥95.0%

Leverantör: TCI EUROPE
Varning

Synonyms: Benzyl 3-bromopropyl ether, 3-bromopropylbenzyl ether

B2818-25G B2818-5G
TCIAB2818-25GEA 3950 SEK
TCIAB2818-25G TCIAB2818-5G
((3-Bromopropoxy)methyl)benzene ≥95.0%
((3-Bromopropoxy)methyl)benzene
Formel: C₁₀H₁₃BrO
MW: 229,117 g/mol
Kokpunkt: 141°C
MDL Number: MFCD00134570
CAS nummer: 54314-84-0

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR