Benzyl-2-bromoethyl ether ≥97.0%
Leverantör: TCI
Varning
B2817-25G B2817-5G
TCIAB2817-25GEA 1500 SEK
TCIAB2817-25G TCIAB2817-5G
Benzyl-2-bromoethyl ether ≥97.0%
Benzyl-2-bromoethyl ether
Formel: C₉H₁₁BrO
Kokpunkt: 95°C
MDL Number: MFCD01321307
CAS nummer: 1462-37-9
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %