Bis(2,6-diisopropylphenyl)carbodiimide ≥98.0%

Leverantör: TCI EUROPE
Fara

Synonyms: N,N'-Methanediylidenebis(2,6-diisopropylaniline)

B2756-25G B2756-5G
TCIAB2756-25GEA 1250 SEK
TCIAB2756-25G TCIAB2756-5G
Bis(2,6-diisopropylphenyl)carbodiimide ≥98.0%
Bis(2,6-diisopropylphenyl)carbodiimide
Formel: C₂₅H₃₄N₂
MW: 362,56 g/mol
MDL Number: MFCD00082211
CAS nummer: 2162-74-5
EINECS: 218-487-5
UN: 2811
ADR: 6.1,III

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Melting point 50.0 to 54.0 deg-C
Solubility in hot Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR