Butylsuccinic anhydride ≥98.0%

Leverantör: TCI
Varning

B2742-25G B2742-5G
TCIAB2742-25GEA 5400 SEK
TCIAB2742-25G TCIAB2742-5G
Butylsuccinic anhydride ≥98.0%
Butylsuccinic anhydride
Formel: C₈H₁₂O₃
Storage Temperature: Kylskåp
CAS nummer: 2035-76-9

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 44.0 to 48.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR