4,4'-Bis(9H-carbazol-9-yl)biphenyl ≥98.0% (by HPLC, total nitrogen)

Leverantör: TCI EUROPE
Fara

Synonyms: CBP, 4,4'-Di(9H-carbazol-9-yl)-1,1'-biphenyl, 4,4-Di(9H-carbazol-9-yl)-1,1-biphenyl

B2713-1G B2713-5G
TCIAB2713-1GEA 845 SEK
TCIAB2713-1G TCIAB2713-5G
4,4'-Bis(9H-carbazol-9-yl)biphenyl ≥98.0% (by HPLC, total nitrogen)
4,4'-Bis(9H-carbazol-9-yl)biphenyl
Formel: C₃₆H₂₄N₂
MW: 484,60 g/mol
Smältpunkt: 283°C
MDL Number: MFCD00093417
CAS nummer: 58328-31-7

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR