2-(4-Bromophenyl)benzothiazole ≥98.0%
Leverantör: TCI EUROPE
Synonyms:
1-(2-Benzothiazolyl)-4-bromobenzene
B2679-25G
B2679-5G
TCIAB2679-25GEA
3125
SEK
TCIAB2679-25G
TCIAB2679-5G
2-(4-Bromophenyl)benzothiazole ≥98.0%
2-(4-Bromophenyl)benzothiazole
Formel:
C₁₃H₈BrNS MW: 290,18 g/mol |
MDL Number:
MFCD00579199 CAS nummer: 19654-19-4 |
Specifikation testresultat
Appearance | White to Light yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 132.0 to 135.0 deg-C |
Solubility in Toluene | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...