1,3,5-Benzenetrithiol ≥98.0%

Leverantör: TCI
Varning

Synonyms: 1,3,5-Trimercaptobenzene

B2656-1G B2656-5G
TCIAB2656-1GEA 3500 SEK
TCIAB2656-1G TCIAB2656-5G
1,3,5-Benzenetrithiol ≥98.0%
1,3,5-Benzenetrithiol
Formel: C₆H₆S₃
MW: 174.29 g/mol
Storage Temperature: Kylskåp
CAS nummer: 38004-59-0

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 58.0 to 62.0 deg-C
Solubility in Acetone almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR