3-Bromoisobutyric acid ≥97.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
Captopril EP Impurity D, 3-Bromo-2-methylpropanoic acid, 3-Bromo-2-methylpropionic Acid
B2571-25G
B2571-5G
TCIAB2571-25GEA
2825
SEK
TCIAB2571-25G
TCIAB2571-5G
3-Bromoisobutyric acid ≥97.0% (by GC, titration analysis)
3-Bromoisobutyric acid
Formel:
C₄H₇BrO₂ MW: 167,00 g/mol |
MDL Number:
MFCD01310530 CAS nummer: 56970-78-6 UN: 3265 ADR: 8,II |
Specifikation testresultat
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 97.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...