4-Benzylthiomorpholine-1,1-dioxide ≥98.0%

Leverantör: TCI EUROPE

B2434-25G B2434-5G
TCIAB2434-25GEA 2075 SEK
TCIAB2434-25G TCIAB2434-5G
4-Benzylthiomorpholine-1,1-dioxide ≥98.0%
4-Benzylthiomorpholine-1,1-dioxide
Formel: C₁₁H₁₅NO₂S
MW: 225,31 g/mol
CAS nummer: 26475-66-1

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Solubility in Acetone almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR