12-Benzylamino-1-dodecanol ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
N-(12-Hydroxydodecyl)benzylamine
B2403-1G
B2403-5G
TCIAB2403-1GEA
910
SEK
TCIAB2403-1G
TCIAB2403-5G
12-Benzylamino-1-dodecanol ≥98.0% (by GC, titration analysis)
12-Benzylamino-1-dodecanol
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Solubility in Methanol | almost transparency |
Melting point | 41.0 to 45.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...