9,9-Bis(4-hydroxy-3-methylphenyl)fluorene ≥98.0%
Leverantör: TCI EUROPE
Synonyms:
4,4'-(9H-Fluorene-9,9-diyl)bis(2-methylphenol), 4,4'-(9-Fluorenylidene)di(o-cresol)
B2396-25G
B2396-250G
TCIAB2396-25GEA
990
SEK
TCIAB2396-25G
TCIAB2396-250G
9,9-Bis(4-hydroxy-3-methylphenyl)fluorene ≥98.0%
9,9-Bis(4-hydroxy-3-methylphenyl)fluorene
Formel:
C₂₇H₂₂O₂ MW: 378,47 g/mol |
MDL Number:
MFCD04038046 CAS nummer: 88938-12-9 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 216.0 to 222.0 deg-C |
Solubility in hot Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...