4-tert-Butoxyphenol ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
Hydroquinone Mono-tert-butyl Ether
B2390-1G
B2390-5G
TCIAB2390-1GEA
815
SEK
TCIAB2390-1G
TCIAB2390-5G
4-tert-Butoxyphenol ≥98.0%
4-tert-Butoxyphenol
Formel:
C₁₀H₁₄O₂ MW: 166,22 g/mol |
MDL Number:
MFCD02183556 CAS nummer: 2460-87-9 |
Specifikation testresultat
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 149.0 to 155.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...