3-Bromo-3,3-difluoropropene 95,0 GC_ANALYSMETOD
Leverantör: TCI
Synonyms: 3,3-Difluoro-3-bromopropene
B2333-1G B2333-5G
TCIAB2333-1GEA 2250 SEK
TCIAB2333-1G TCIAB2333-5G
3-Bromo-3,3-difluoropropene 95,0 GC_ANALYSMETOD
3-Bromo-3,3-difluoropropene
Formel: C₃H₃BrF₂
MW: 156.96 g/mol
Kokpunkt: 42°C
Storage Temperature: Kylskåp
MDL Number: MFCD00042185
CAS nummer: 420-90-6
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %