Ethyl-9-bromononanoate ≥95.0%

Leverantör: TCI
Varning

Synonyms: 9-Bromononanoic Acid Ethyl Ester, Ethyl 9-bromononanoate

B2324-1G B2324-5G
TCIAB2324-1GEA 1725 SEK
TCIAB2324-1G TCIAB2324-5G
Ethyl-9-bromononanoate ≥95.0%
Ethyl-9-bromononanoate
Formel: C₁₁H₂₁BrO₂
Kokpunkt: 180°C
MDL Number: MFCD06797065
CAS nummer: 28598-81-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9dbb3b90de863cac5e62b0f45d606c113a806b95&vid=9dbb3b90de863cac5e62b0f45d606c113a806b95&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR