2-[4-(tert-Butyldimethylsilyloxymethyl)-2-nitrophenoxy]acetic acid ≥98.0% (by HPLC, titration analysis)

Leverantör: TCI EUROPE

B2322-1G B2322-100MG
TCIAB2322-1GEA 6925 SEK
TCIAB2322-1G TCIAB2322-100MG
2-[4-(tert-Butyldimethylsilyloxymethyl)-2-nitrophenoxy]acetic acid ≥98.0% (by HPLC, titration analysis)
2-[4-(tert-Butyldimethylsilyloxymethyl)-2-nitrophenoxy]acetic acid
Formel: C₁₅H₂₃NO₆Si
MDL Number: MFCD04117911
CAS nummer: 308815-83-0

Order Now

Specifikation testresultat

Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR