tert-Butyl (2-bromoethyl)carbamate ≥98.0%

Leverantör: TCI
Varning

Synonyms: 2-(BOC-amino)ethyl bromide

B2289-1G B2289-25G B2289-5G
TCIAB2289-1GEA 565 SEK
TCIAB2289-1G TCIAB2289-25G TCIAB2289-5G
tert-Butyl (2-bromoethyl)carbamate ≥98.0%
tert-Butyl (2-bromoethyl)carbamate
Formel: C₇H₁₄BrNO₂
MW: 224.10 g/mol
Storage Temperature: Frys
MDL Number: MFCD02683428
CAS nummer: 39684-80-5

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR