4-Bromobenzene-1,2-diol ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 4-Bromocatechol, 4-Bromo-1,2-dihydroxybenzene

B2173.0005 B2173-1G B2173-5G
TCIAB2173.0005EA 5675 SEK
TCIAB2173.0005 TCIAB2173-1G TCIAB2173-5G
4-Bromobenzene-1,2-diol ≥98.0%
4-Bromobenzene-1,2-diol
Formel: C₆H₅BrO₂
MW: 189,0067 g/mol
MDL Number: MFCD00869769
CAS nummer: 17345-77-6
EINECS: 241-370-5

Order Now

Specifikation testresultat

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 83.0 to 87.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR