Ethyl 2-(2-Benzothiazolyl)acetate ≥95.0%
Leverantör: TCI EUROPE
Synonyms:
2-Benzothiazoleacetic acid Ethyl ester, Ethyl 2-Benzothiazoleacetate, 2-(2-Benzothiazolyl)acetic acid Ethyl ester
B2168-5G
B2168-1G
TCIAB2168-5GEA
1525
SEK
TCIAB2168-5G
TCIAB2168-1G
Ethyl 2-(2-Benzothiazolyl)acetate ≥95.0%
Ethyl 2-(2-Benzothiazolyl)acetate
Formel:
C₁₁H₁₁NO₂S MW: 221,27 g/mol Kokpunkt: 145°C Storage Temperature: Kylskåp |
MDL Number:
MFCD00546397 CAS nummer: 29182-42-1 |
Specifikation testresultat
Appearance | Light yellow to Yellow to Orange clear liquid |
Purity (GC) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...