4,4'-Bis(chloromethyl)biphenyl ≥95.0%
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
4,4-Bis(chloromethyl)-1,1-biphenyl, 4,4'-Bis(chloromethyl)-1,1-biphenyl
B2099-25G
B2099-500G
TCIAB2099-25GEA
215
SEK
TCIAB2099-25G
TCIAB2099-500G
4,4'-Bis(chloromethyl)biphenyl ≥95.0%
4,4'-Bis(chloromethyl)biphenyl
Formel:
C₁₄H₁₂Cl₂ MW: 251,15 g/mol Kokpunkt: 184°C |
MDL Number:
MFCD00674019 CAS nummer: 1667-10-3 EINECS: 216-784-4 UN: 3261 ADR: 8,II |
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 95.0 % |
Melting point | 139.0 to 143.0 deg-C |