Methyl-3,5-dibenzyloxybenzoate ≥98.0%

Leverantör: TCI EUROPE

Synonyms: 3,5-Bis(benzyloxy)benzoic Acid Methyl Ester, 3,5-Dibenzyloxybenzoic Acid Methyl Ester

B2092-25G B2092-5G
TCIAB2092-25GEA 3425 SEK
TCIAB2092-25G TCIAB2092-5G
Methyl-3,5-dibenzyloxybenzoate ≥98.0%
Methyl-3,5-dibenzyloxybenzoate
Formel: C₂₂H₂₀O₄
MW: 348,40 g/mol
MDL Number: MFCD00157205
CAS nummer: 58605-10-0

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 69.0 to 72.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR