N-ɑ-Boc-N(im)-tosyl-L-histidine ≥98.0%

Leverantör: TCI EUROPE

Synonyms: N-Boc-1-(p-toluenesulphonyl)-L-histidine, (S)-2-((tert-Butoxycarbonyl)amino)-3-(1-tosyl-1H-imidazol-4-yl)propanoic acid, N-ɑ-(tert-Butoxycarbonyl)-tele-(p-toluenesulphonyl)-L-histidine, Boc-His(Tos)-OH

B2066-25G B2066-5G
TCIAB2066-25GEA 3550 SEK
TCIAB2066-25G TCIAB2066-5G
N-ɑ-Boc-N(im)-tosyl-L-histidine ≥98.0%
N-alpha-Boc-N(im)-tosyl-L-histidine
Formel: C₁₈H₂₃N₃O₆S
MW: 409,46 g/mol
MDL Number: MFCD00065967
CAS nummer: 35899-43-5

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D +13.0 to +16.0 deg (C=1, MeOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR