(1S,4S)-2-Benzyl-2,5-diazabicyclo[2.2.1]heptane dihydrobromide ≥98.0%

Leverantör: TCI EUROPE
Varning

B2054-1G B2054-5G
TCIAB2054-1GEA 1475 SEK
TCIAB2054-1G TCIAB2054-5G
(1S,4S)-2-Benzyl-2,5-diazabicyclo[2.2.1]heptane dihydrobromide ≥98.0%
(1S,4S)-2-Benzyl-2,5-diazabicyclo[2.2.1]heptane dihydrobromide
Formel: C₁₂H₁₆N₂
MW: 350,10 g/mol
MDL Number: MFCD01321292
CAS nummer: 116258-17-4

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (Argentmetric Titration) min. 98.0 %
Specific rotation [a]20/D +15.0 to +17.0 deg (C=3, 2mol/L NaOH sol.)
Solubility in Water very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR